Learn More
Sino Biological Recombinant Rat Her2/ERBB2 Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 80079R08H100
Encompass_Preferred
A DNA sequence encoding the rat ErbB2 (P06494) extracellular domain (Met1-Thr654) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3669