Learn More
Sino Biological Recombinant Rat Her2/ERBB2 Protein
Supplier:  Sino Biological 80079RCCH50
Encompass_Preferred
A DNA sequence encoding the rat ErbB2 (P06494) extracellular domain (Met1-Thr654) was expressed and purified, with seven additional amino acids (ALEVLFQ) at the C-terminus.
Catalog No. 50-162-3672