Learn More
Sino Biological Recombinant Rat IL-7R alpha/CD127 Protein (mFc Tag)
Supplier:  Sino Biological 80083R05H50
Encompass_Preferred
A DNA sequence encoding the rat IL7R (D4A3X8) (Met1-Asp239) was fused with Fc region of mouse IgG1 at the C-terminus.
Catalog No. 50-162-3676