Learn More
Sino Biological Recombinant Rat IL-7R alpha/CD127 Protein (His Tag)
Supplier: Sino Biological 80083R08H100
A DNA sequence encoding the rat IL7R (NP_001099888.1) (Met1-Asp239) was expressed with a polyhistidine tag at the C-terminus.