Learn More
Sino Biological Recombinant Rat UCHL3 Protein (His Tag)
Supplier:  Sino Biological 80087R07E50
Encompass_Preferred
A DNA sequence encoding the rat UCHL3 (Q91Y78) (Glu 2-Ala 230) was fused with a polyhistidine tag at the N-terminus.
Catalog No. 50-162-3682