Learn More
Sino Biological Recombinant Rat FGFR4 Protein (hFc Tag)
Supplier:  Sino Biological 80093R02H100
Encompass_Preferred
A DNA sequence encoding the rat FGFR4 (Q498D6) extracellular domain (Met 1-Asp 367) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-3685