Learn More
Sino Biological Recombinant Rat FGFR4 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 80093R08H100
Encompass_Preferred
A DNA sequence encoding the rat FGFR4 (Q498D6) (Met1-Asp367) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3687