Learn More
Alkali Scientific Alkali Scientific™ 800uL RAININ Compatible Black Conductive Robotic Automation Tips , 96-Well Head/Tip Format, Low Retention, Sterile, Filtered, 960/Pack
SDP
Supplier:  Alkali Scientific RFT0800
Encompass
Enhance your robotic liquid handling with our 800µL Rainin Compatible Black Conductive Tips. Designed for 96-well head/tip formats, these low-retention tips ensure precise and consistent liquid transfers for automated systems. Ideal for larger volume applications, these sterilized, racked conductive filter tips are fully compatible with Rainin LTS™ nozzles, offering seamless integration into your robotic liquid handling systems. The tips are packaged in a standard Rainin™ robotic tip rack featuring an SBS 8x12 tip deck format, making organization and access effortless. Certified free from DNA, DNase, RNase, ATP, pyrogens, PCR inhibitors, additives, and bioactive contaminants, our tips guarantee cleanliness for contamination-free handling. Improve your automation efficiency with these high-performance tips, delivering exceptional accuracy in your lab. Not compatible with Rainin BenchSmart 96™ or Liquidator 96™.
Catalog No. 50-302-8344