Learn More
Sino Biological Mouse / Rat VEGFC / VEGF-C Protein (aa 108-223, His Tag)
Supplier:  Sino Biological 80103R08H50
Encompass_Preferred
A DNA sequence encoding the mouse / rat VEGFC (NP_033532.1 & O35757) (Ala108-Arg223) was expressed fused with a polyhistidine tag at the C-terminus. Mouse and Rat mature VEGFC sequences are identical.
Catalog No. 50-162-3698