Learn More
Sino Biological Recombinant Rat VEGF-D Protein (hFc Tag)
Supplier:  Sino Biological 80104R01H50
Encompass_Preferred
A DNA sequence encoding the rat FIGF (NP_113949.1)(Phe94-Arg210) was expressed, fused with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-162-3700