Learn More
Sino Biological Recombinant Rat Ephrin A5 Protein (His Tag)
Supplier: Sino Biological 80105R08H100
A DNA sequence encoding the rat EFNA5 (P97605) (Met 1-Glu 202), without the pro peptide, was expressed, fused with a polyhistidine tag at the C-terminus.