Learn More
Sino Biological Recombinant Rat Ephrin-B1 Protein (His Tag)
Supplier:  Sino Biological 80106R08H200
Encompass_Preferred
A DNA sequence encoding the rat EFNB1 (P52796) extracellular domain (Met 1-Thr 229) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3708