Learn More
Sino Biological Recombinant Rat Ephrin B2 Protein (hFc Tag)
Supplier:  Sino Biological 80107R02H100
Encompass_Preferred
A DNA sequence encoding the rat EFNB2 (NP_001100798.1) extracellular domain (Met 1-Ala 229) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-3709