Learn More
Sino Biological Recombinant Rat Ephrin B2 Protein (His Tag)
Supplier:  Sino Biological 80107R08H200
Encompass_Preferred
A DNA sequence encoding the rat EFNB2 (NP_001100798.1) (Met1-Ala229) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3712