Learn More
Sino Biological Recombinant Rat EphA7 Protein (hFc Tag)
Supplier:  Sino Biological 80109R02H200
Encompass_Preferred
A DNA sequence encoding the rat Epha7 (NP_599158.1) (Met1-Ser539) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-3714