Learn More
Sino Biological Recombinant Rat EphA7 Protein (His Tag)
Supplier:  Sino Biological 80109R08H200
Encompass_Preferred
A DNA sequence encoding the rat EPHA7 (Met1-Ser539) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3716