Learn More
Sino Biological Recombinant Rat Ephrin A2/EFNA2 Protein (hFc Tag)
Supplier:  Sino Biological 80110R02H200
Encompass_Preferred
A DNA sequence encoding the rat Efna2 (NP_001162141.1) (Met1-Ser183) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-3718