Learn More
Sino Biological Recombinant Rat Ephrin B3 Protein (hFc Tag)
Supplier:  Sino Biological 80112R02H100
Encompass_Preferred
A DNA sequence encoding the rat EFNB3 (NP_001094450) (Met1-Ser224) was expressed, fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-3724