Learn More
Sino Biological Recombinant Rat Ephrin B3 Protein (His Tag)
Supplier: Sino Biological 80112R08H100
A DNA sequence encoding the rat EFNB3 (G3V7D4) (Met1-Ser224) was expressed, fused with a polyhistidine tag at the C-terminus.