Learn More
Sino Biological Recombinant Rat VEGFR2/KDR Protein (hFc Tag)
Supplier:  Sino Biological 80113R02H100
Encompass_Preferred
A DNA sequence encoding the rat KDR (AAH87029.1) (Met1-Glu760) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-3728