Learn More
Sino Biological Recombinant Rat Ephrin-A1 Protein (His Tag)
Supplier:  Sino Biological 80115R08H200
Encompass_Preferred
A DNA sequence encoding the rat EFNA1 (P97553) (Met1-His181) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3732