Learn More
Sino Biological Recombinant Rat TGF beta 1 Protein (Latent, His Tag)
Supplier:  Sino Biological 80116R08H20
Encompass_Preferred
A DNA sequence encoding the rat Latent TGFB1 (P17246) (Met1-Ser390) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3735