Learn More
Sino Biological TGF beta 1 Protein, Rat/Mouse, Recombinant
Supplier:  Sino Biological 80116RNAH5
Encompass_Preferred
A DNA sequence encoding the mouse / rat TGFB1 (Ala279-Ser390) was expressed. Rat and Mouse mature TGFB1 sequences are identical.
Catalog No. 50-162-3737