Learn More
Sino Biological Recombinant Rat ACVR1 Protein (hFc Tag)
Supplier:  Sino Biological 80118R02H200
Encompass_Preferred
A DNA sequence encoding the rat ACVR1 (P80201) (Met1-Glu123) was expressed, fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-3739