Learn More
Sino Biological Recombinant Rat EphA4 Protein (hFc Tag)
Supplier: Sino Biological 80123R02H200
A DNA sequence encoding the rat EPHA4 (D3ZZK3) (Met1-Tyr547) was expressed, fused with the Fc region of human IgG1 at the C-terminus.