Learn More
Sino Biological Recombinant Rat EphA4 Protein (His Tag)
Supplier:  Sino Biological 80123R08H200
Encompass_Preferred
A DNA sequence encoding the rat EPHA4 (D3ZZK3) (Met1-Thr547) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3742