Learn More
Sino Biological Recombinant Rat ERBB4/Her4 Protein (hFc Tag)
Supplier: Sino Biological 80125R02H100
A DNA sequence encoding the rat ERBB4 (AAQ77349.1)(Met1-Pro651) was expressed with the Fc region of human IgG1 at the C-terminus.