Learn More
Sino Biological Recombinant Rat CD30 Ligand/TNFSF8 Protein (hFc Tag)
Supplier:  Sino Biological 80148R01H100
Encompass_Preferred
A DNA sequence encoding the rat TNFSF8 (NP_001305962.1)(Gln66-Asp237) was expressed with Fc region of human IgG1 at the N-terminus.
Catalog No. 50-162-3756