Learn More
Sino Biological Recombinant Rat CD30 Ligand/TNFSF8 Protein
Supplier:  Sino Biological 80148RNCH100
Encompass_Preferred
A DNA sequence encoding the rat TNFSF8 (NP_001305962.1)(Gln66-Asp237) was expressed with two additional amino acids (Gly & Pro) at the N-terminus.
Catalog No. 50-162-3758