Learn More
Sino Biological Recombinant Rat CXCL16 Protein (hFc Tag)
Supplier:  Sino Biological 80149R02H50
Encompass_Preferred
A DNA sequence encoding the rat CXCL16 (NP_001017478.1)(Met1-Ala198) was expressed, fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-3761