Learn More
Sino Biological Recombinant Rat CXCL16 Protein (His Tag)
Supplier:  Sino Biological 80149R08H50
Encompass_Preferred
A DNA sequence encoding the rat CXCL16 (Q6AXU5) extracellular domain (Met 1-Ala 198) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3763