Learn More
Sino Biological Recombinant Rat CD40/TNFRSF5 Protein (hFc Tag)
Supplier:  Sino Biological 80151R02H100
Encompass_Preferred
A DNA sequence encoding the rat CD40 (Q4QQW2) (Met1-Arg193) was expressed, fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-3767