Learn More
Sino Biological Recombinant Rat CD40/TNFRSF5 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 80151R08H100
Encompass_Preferred
A DNA sequence encoding the rat CD40 (NP_599187.1)(Met1-Arg193) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3769