Learn More
Sino Biological Recombinant Rat TWEAK/TNFSF12 Protein (hFc Tag)
Supplier:  Sino Biological 80154R01H50
Encompass_Preferred
A DNA sequence encoding the rat TNFSF12 (NP_001001513.2)(Arg105-His249) was expressed with Fc region of human IgG1 at the N-terminus.
Catalog No. 50-162-3773