Learn More
Sino Biological Recombinant Rat CD2 Protein (hFc Tag)
Supplier:  Sino Biological 80158R02H100
Encompass_Preferred
A DNA sequence encoding the rat CD2 (Q5I0M6) (Met1-Pro202) was expressed, fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-3778