Learn More
Sino Biological Recombinant Rat RANK/TNFRSF11A Protein (hFc Tag)
Supplier:  Sino Biological 80160R02H100
Encompass_Preferred
A DNA sequence encoding the rat TNFRSF11A (NP_001258164.1)(Met1-Pro213) was expressed, fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-3780