Learn More
Sino Biological Recombinant Rat RANK/TNFRSF11A Protein (His Tag)
Supplier:  Sino Biological 80160R08H100
Encompass_Preferred
A DNA sequence encoding the rat TNFRSF11A (XP_573424.2) extracellular domain (Met 1-Pro 213) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3782