Learn More
Sino Biological Recombinant Rat CD70/CD27 Ligand Protein (hFc Tag)
Supplier: Sino Biological 80161R01H100
A DNA sequence encoding the rat CD70 (D3ZGU3) (Gln46-Pro195) was expressed, fused with Fc region of human IgG1 at the N-terminus.