Learn More
Sino Biological Recombinant Rat 4-1BB Ligand/TNFSF9 Protein (hFc Tag)
Supplier: Sino Biological 80166R01H50
A DNA sequence encoding the rat TNFSF9 (Q80WE6) (Pro106-Gly308) was expressed, fused with Fc region of human IgG1 at the N-terminus.