Learn More
Sino Biological Recombinant Rat 4-1BB Ligand/TNFSF9 Protein (His Tag)
Supplier:  Sino Biological 80166R07H50
Encompass_Preferred
A DNA sequence encoding the rat TNFSF9 (Q80WE6) (Pro106-Gly308) was expressed, fused with a polyhistidine tag at the N-terminus.
Catalog No. 50-162-3791