Learn More
Sino Biological Recombinant Rat BAFFR/TNFRSF13C Protein (hFc Tag)
Supplier: Sino Biological 80171R02H100
A DNA sequence encoding the rat TNFRSF13C (XP_576316.2) (Ser10-Ala71) was expressed with the Fc region of human IgG1 at the C-terminus.