Learn More
Sino Biological Recombinant Rat Interferon alpha 4/IFNA4 Protein (hFc Tag)
Supplier:  Sino Biological 80173R01H100
Encompass_Preferred
A DNA sequence encoding the rat IFNA4 (D3ZFH0) (Cys24-Lys189) was expressed, fused with Fc region of human IgG1 at the N-terminus.
Catalog No. 50-162-3796