Learn More
Sino Biological Recombinant Rat Interferon alpha 1/IFNA1 Protein (His Tag)
Supplier:  Sino Biological 80174R08H100
Encompass_Preferred
A DNA sequence encoding the rat IFNA1 (P05011) (Met1-Ser192) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3801