Learn More
Sino Biological Recombinant Rat Lymphotoxin-beta receptor/TNFRSF3 Protein (hFc Tag)
Supplier:  Sino Biological 80178R02H100
Encompass_Preferred
A DNA sequence encoding the rat LTBR (Q5U2S8) (Met1-Ala218) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-3806