Learn More
Sino Biological Recombinant Rat Lymphotoxin-beta receptor/TNFRSF3 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 80178R08H100
Encompass_Preferred
A DNA sequence encoding the rat LTBR (NP_001008316.1) extracellular domain (Met 1-Ala 218) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3808