Learn More
Sino Biological Recombinant Rat Il11ra1 Protein (His Tag)
Supplier:  Sino Biological 80179R08H200
Encompass_Preferred
A DNA sequence encoding the Rat Il11ra1 (NP_620816.1) (Met1-Gln366) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3810