Learn More
Sino Biological Recombinant Rat TNFR1/CD120a Protein (hFc Tag)
Supplier:  Sino Biological 80181R02H100
Encompass_Preferred
A DNA sequence encoding the rat TNFRSF1A (P22934) (Met1-Ala211) was expressed, fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-3812