Learn More
Sino Biological Recombinant Rat TNFR1/CD120a Protein (His Tag)
Supplier:  Sino Biological 80181R08H100
Encompass_Preferred
A DNA sequence encoding the rat TNFRSF1A (NP_037223.1)(Met1-Ala211) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3814