Learn More
Sino Biological Recombinant Rat IFNGR1 Protein (hFc Tag)
Supplier: Sino Biological 80182R02H100
A DNA sequence encoding the rat IFNGR (Q6P6T3) (Met1-Ser241) was expressed with the Fc region of human IgG1 at the C-terminus.