Learn More
Sino Biological Recombinant Rat IL-13 R alpha 1/IL13RA1 Protein (hFc Tag)
Supplier:  Sino Biological 80186R02H100
Encompass_Preferred
A DNA sequence encoding the rat IL13RA1 (Q561K3) (Met1-Thr342) was expressed, fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-3821